Assay Details
| Gene Symbol: LOC4336300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780153.2 | XP_015635639.1 | 7 - 8 | 1284 | 103 | |
| GenBank mRNA | AK065553.1 | - | 8 - 9 | 1116 | 103 | |
| AY224450.1 | - | 7 - 8 | 1072 | 103 | ||
More Information
Set Membership: |
|

LOC4336300
Genomic Map