Assay Details
| Gene Symbol: LOC4336300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780153.2 | XP_015635639.1 | 8 - 9 | 1531 | 113 | |
| GenBank mRNA | AK060761.1 | - | 2 - 3 | 269 | 113 | |
| AK065553.1 | - | 9 - 10 | 1363 | 113 | ||
| AY224450.1 | - | 8 - 9 | 1319 | 113 | ||
More Information
Set Membership: |
|

LOC4336300
Genomic Map