Assay Details
| Gene Symbol: LOC4336983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781223.1 | XP_015636709.1 | 16 - 17 | 1461 | 69 | |
| GenBank mRNA | AF251073.1 | - | 6 - 7 | 425 | 69 | |
| AK061967.1 | - | 12 - 13 | 879 | 69 | ||
| AK102306.1 | - | 16 - 17 | 1462 | 69 | ||
More Information
Set Membership: |
|

LOC4336983
Genomic Map