Assay Details
| Gene Symbol: LOC4336983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781223.1 | XP_015636709.1 | 18 - 19 | 1661 | 105 | |
| GenBank mRNA | AF251073.1 | - | 8 - 9 | 625 | 105 | |
| AK061967.1 | - | 14 - 15 | 1079 | 105 | ||
| AK102306.1 | - | 18 - 19 | 1662 | 105 | ||
More Information
Set Membership: |
|

LOC4336983
Genomic Map