Assay Details
| Gene Symbol: LOC4336983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781223.1 | XP_015636709.1 | 1 - 2 | 234 | 73 | |
| GenBank mRNA | AK102306.1 | - | 1 - 2 | 235 | 73 | |
More Information
Set Membership: |
|

LOC4336983
Genomic Map