Assay Details
| Gene Symbol: LOC4336983 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781223.1 | XP_015636709.1 | 2 - 3 | 365 | 98 | |
| GenBank mRNA | AK102306.1 | - | 2 - 3 | 366 | 98 | |
More Information
Set Membership: |
|

LOC4336983
Genomic Map