Assay Details
| Gene Symbol: LOC4337057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778835.1 | XP_015634321.1 | 4 - 5 | 411 | 67 | |
| XM_015778838.2 | XP_015634324.1 | 4 - 5 | 411 | 67 | ||
| GenBank mRNA | AK070522.1 | - | 4 - 5 | 348 | 67 | |
More Information
Set Membership: |
|

LOC4337057
Genomic Map