Assay Details
Gene Symbol: LOC4337651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782106.2 | XP_015637592.1 | 3 - 4 | 530 | 121 | |
XM_015782107.2 | XP_015637593.1 | 3 - 4 | 530 | 121 | ||
GenBank mRNA | AK066388.1 | - | 3 - 4 | 488 | 121 | |
AK120088.1 | - | 3 - 4 | 486 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
