Assay Details
Gene Symbol: LOC4337651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782106.2 | XP_015637592.1 | 6 - 7 | 759 | 109 | |
XM_015782107.2 | XP_015637593.1 | 6 - 7 | 759 | 109 | ||
GenBank mRNA | AK066388.1 | - | 6 - 7 | 717 | 109 | |
AK120088.1 | - | 6 - 7 | 715 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
