Assay Details
| Gene Symbol: LOC4338353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784554.2 | XP_015640040.1 | 1 - 2 | 125 | 65 | |
| GenBank mRNA | AK061642.1 | - | 1 - 2 | 117 | 65 | |
| AK103134.1 | - | 1 - 2 | 120 | 65 | ||
| AK103615.1 | - | 1 - 2 | 124 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015784554.2 transcripts.
Set Membership: |
|

LOC4338353
Genomic Map