Assay Details
| Gene Symbol: LOC4338353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784554.2 | XP_015640040.1 | 2 - 3 | 618 | 76 | |
| GenBank mRNA | AK061642.1 | - | 2 - 3 | 610 | 76 | |
| AK103134.1 | - | 2 - 3 | 613 | 76 | ||
| AK103615.1 | - | 2 - 3 | 617 | 76 | ||
More Information
Set Membership: |
|

LOC4338353
Genomic Map