Assay Details
| Gene Symbol: LOC4338353 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784554.2 | XP_015640040.1 | 3 - 4 | 988 | 75 | |
| GenBank mRNA | AK061642.1 | - | 3 - 4 | 980 | 75 | |
| AK103134.1 | - | 3 - 4 | 983 | 75 | ||
| AK103615.1 | - | 3 - 4 | 987 | 75 | ||
More Information
Set Membership: |
|

LOC4338353
Genomic Map