Assay Details
Gene Symbol: LOC4339406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782182.2 | XP_015637668.1 | 5 - 6 | 834 | 67 | |
XM_015782183.2 | XP_015637669.1 | 5 - 6 | 831 | 67 | ||
GenBank mRNA | AK062298.1 | - | 5 - 6 | 793 | 67 | |
AK064168.1 | - | 5 - 6 | 792 | 67 | ||
AK108715.1 | - | 5 - 6 | 787 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
