Assay Details
| Gene Symbol: LOC4341475 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785637.2 | XP_015641123.1 | 3 - 4 | 1376 | 146 | |
| GenBank mRNA | AB212075.1 | - | 6 - 7 | 1731 | 146 | |
| AK106613.1 | - | 1 - 2 | 716 | 146 | ||
More Information
Set Membership: |
|

LOC4341475
Genomic Map