Assay Details
| Gene Symbol: LOC4342150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787505.2 | XP_015642991.1 | 1 - 2 | 275 | 108 | |
| GenBank mRNA | AK062118.1 | - | 1 - 2 | 160 | 108 | |
| AK064963.1 | - | 1 - 2 | 264 | 108 | ||
| AK065172.1 | - | 1 - 2 | 210 | 108 | ||
| AK099361.1 | - | 1 - 2 | 213 | 108 | ||
More Information
Set Membership: |
|

LOC4342150
Genomic Map