Assay Details
| Gene Symbol: LOC4342150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787505.2 | XP_015642991.1 | 2 - 3 | 433 | 81 | |
| GenBank mRNA | AK062118.1 | - | 2 - 3 | 318 | 81 | |
| AK065172.1 | - | 2 - 3 | 368 | 81 | ||
| AK099361.1 | - | 2 - 3 | 371 | 81 | ||
More Information
Set Membership: |
|

LOC4342150
Genomic Map