Assay Details
| Gene Symbol: LOC4342150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787505.2 | XP_015642991.1 | 4 - 5 | 657 | 71 | |
| GenBank mRNA | AK062118.1 | - | 4 - 5 | 542 | 71 | |
| AK064963.1 | - | 4 - 5 | 646 | 71 | ||
| AK065172.1 | - | 4 - 5 | 592 | 71 | ||
| AK099361.1 | - | 4 - 5 | 595 | 71 | ||
More Information
Set Membership: |
|

LOC4342150
Genomic Map