Assay Details
| Gene Symbol: LOC4342743 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791285.2 | XP_015646771.1 | 1 - 2 | 320 | 63 | |
| GenBank mRNA | AK060442.1 | - | 1 - 2 | 280 | 63 | |
| AK100823.1 | - | 1 - 2 | 305 | 63 | ||
| AK103131.1 | - | 1 - 2 | 278 | 63 | ||
| AY224456.1 | - | 1 - 2 | 222 | 63 | ||
More Information
Set Membership: |
|

LOC4342743
Genomic Map