Assay Details
| Gene Symbol: LOC4343420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792402.2 | XP_015647888.1 | 10 - 11 | 1009 | 86 | |
| XM_015792403.2 | XP_015647889.1 | 10 - 11 | 1003 | 86 | ||
| GenBank mRNA | AK099660.1 | - | 10 - 11 | 1004 | 86 | |
More Information
Set Membership: |
|

LOC4343420
Genomic Map