Assay Details
| Gene Symbol: LOC4343420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792402.2 | XP_015647888.1 | 13 - 14 | 1546 | 133 | |
| XM_015792403.2 | XP_015647889.1 | 13 - 14 | 1540 | 133 | ||
| GenBank mRNA | AK099660.1 | - | 13 - 14 | 1541 | 133 | |
More Information
Set Membership: |
|

LOC4343420
Genomic Map