Assay Details
| Gene Symbol: LOC4343420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792402.2 | XP_015647888.1 | 1 - 2 | 286 | 72 | |
| XM_015792403.2 | XP_015647889.1 | 1 - 2 | 285 | 72 | ||
| GenBank mRNA | AK099660.1 | - | 1 - 2 | 281 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792403.2 transcripts.
Set Membership: |
|

LOC4343420
Genomic Map