Assay Details
| Gene Symbol: LOC4343612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791776.2 | XP_015647262.1 | 10 - 11 | 1550 | 84 | |
| GenBank mRNA | AK059690.1 | - | 6 - 7 | 963 | 84 | |
| AK067125.1 | - | 10 - 11 | 1323 | 84 | ||
| AK068892.1 | - | 10 - 11 | 1557 | 84 | ||
| EU847007.1 | - | 10 - 11 | 1298 | 84 | ||
More Information
Set Membership: |
|

LOC4343612
Genomic Map