Assay Details
| Gene Symbol: LOC4343612 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791776.2 | XP_015647262.1 | 8 - 9 | 1223 | 134 | |
| GenBank mRNA | AK059690.1 | - | 4 - 5 | 636 | 134 | |
| AK067125.1 | - | 8 - 9 | 996 | 134 | ||
| AK067240.1 | - | 8 - 9 | 1231 | 134 | ||
| AK068892.1 | - | 8 - 9 | 1230 | 134 | ||
| EU847007.1 | - | 8 - 9 | 971 | 134 | ||
More Information
Set Membership: |
|

LOC4343612
Genomic Map