Assay Details
| Gene Symbol: LOC9266372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794706.2 | XP_015650192.1 | 2 - 3 | 530 | 107 | |
| GenBank mRNA | AK072710.1 | - | 2 - 3 | 524 | 107 | |
More Information
Set Membership: |
|

LOC9266372
Genomic Map