Assay Details
| Gene Symbol: LOC9266372 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794706.2 | XP_015650192.1 | 7 - 8 | 1585 | 118 | |
| GenBank mRNA | AK072710.1 | - | 6 - 7 | 1655 | 118 | |
More Information
Set Membership: |
|

LOC9266372
Genomic Map