Assay Details
| Gene Symbol: LOC4345819 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794751.2 | XP_015650237.1 | 3 - 4 | 1008 | 69 | |
| XM_015794752.2 | XP_015650238.1 | 3 - 4 | 1004 | 69 | ||
More Information
Set Membership: |
|

LOC4345819
Genomic Map