Assay Details
| Gene Symbol: LOC4347546 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015757103.2 | XP_015612589.1 | 11 - 12 | 1290 | 106 | |
| GenBank mRNA | AK101420.1 | - | 11 - 12 | 1238 | 106 | |
More Information
Set Membership: |
|

LOC4347546
Genomic Map