Assay Details
| Gene Symbol: LOC4347658 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015756556.2 | XP_015612042.1 | 1 | 1272 | 72 | |
| GenBank mRNA | AK121655.1 | - | 1 | 1246 | 72 | |
More Information
Set Membership: |
|

LOC4347658
Genomic Map