Assay Details
| Gene Symbol: LOC4330966 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767072.2 | XP_015622558.1 | 10 - 11 | 1665 | 62 | |
| GenBank mRNA | AK060825.1 | - | 6 - 7 | 685 | 62 | |
| AK066747.1 | - | 9 - 10 | 1754 | 62 | ||
More Information
Set Membership: |
|

LOC4330966
Genomic Map