Assay Details
| Gene Symbol: LOC4330995 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767978.2 | XP_015623464.1 | 6 - 7 | 628 | 79 | |
| XM_015767979.2 | XP_015623465.1 | 6 - 7 | 630 | 79 | ||
| GenBank mRNA | AK065845.1 | - | 7 - 8 | 338 | 79 | |
More Information
Set Membership: |
|

LOC4330995
Genomic Map