Assay Details
| Gene Symbol: LOC4334172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777686.2 | 1 - 2 | 201 | 60 | ||
| GenBank mRNA | AK067626.1 | - | 1 - 2 | 188 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4334172| Gene Symbol: LOC4334172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777686.2 | 1 - 2 | 201 | 60 | ||
| GenBank mRNA | AK067626.1 | - | 1 - 2 | 188 | 60 | |
Set Membership: |
|