Assay Details
| Gene Symbol: LOC4334172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777686.2 | 2 - 3 | 875 | 109 | ||
| GenBank mRNA | AK067626.1 | - | 3 - 4 | 626 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4334172| Gene Symbol: LOC4334172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777686.2 | 2 - 3 | 875 | 109 | ||
| GenBank mRNA | AK067626.1 | - | 3 - 4 | 626 | 109 | |
Set Membership: |
|