Assay Details
| Gene Symbol: LOC4334334 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774345.2 | XP_015629831.1 | 9 - 10 | 867 | 108 | |
| XM_015774348.2 | XP_015629834.1 | 9 - 10 | 819 | 108 | ||
| XM_015774349.2 | XP_015629835.1 | 9 - 10 | 811 | 108 | ||
| GenBank mRNA | AK288769.1 | - | 9 - 10 | 868 | 108 | |
More Information
Set Membership: |
|

LOC4334334
Genomic Map