Assay Details
| Gene Symbol: LOC4334891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780877.2 | XP_015636363.1 | 3 - 4 | 439 | 85 | |
| XM_015780878.2 | XP_015636364.1 | 3 - 4 | 373 | 85 | ||
| GenBank mRNA | AK242581.1 | - | 2 - 3 | 596 | 85 | |
More Information
Set Membership: |
|

LOC4334891
Genomic Map