Assay Details
| Gene Symbol: LOC4334891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780877.2 | XP_015636363.1 | 7 - 8 | 836 | 125 | |
| XM_015780878.2 | XP_015636364.1 | 7 - 8 | 770 | 125 | ||
| GenBank mRNA | AK242581.1 | - | 6 - 7 | 994 | 125 | |
More Information
Set Membership: |
|

LOC4334891
Genomic Map