Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 12 - 13 | 1145 | 66 | |
| XM_015781018.2 | XP_015636504.1 | 12 - 13 | 1044 | 66 | ||
| XM_015781019.2 | XP_015636505.1 | 10 - 11 | 830 | 66 | ||
| XM_015781021.2 | XP_015636507.1 | 11 - 12 | 910 | 66 | ||
| GenBank mRNA | AK060337.1 | - | 6 - 7 | 385 | 66 | |
| AK069000.1 | - | 12 - 13 | 1076 | 66 | ||
More Information
Set Membership: |
|

LOC4335342
Genomic Map