Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 14 - 15 | 1315 | 123 | |
| XM_015781018.2 | XP_015636504.1 | 14 - 15 | 1214 | 123 | ||
| XM_015781019.2 | XP_015636505.1 | 12 - 13 | 1000 | 123 | ||
| XM_015781021.2 | XP_015636507.1 | 13 - 14 | 1080 | 123 | ||
| GenBank mRNA | AK060337.1 | - | 8 - 9 | 555 | 123 | |
| AK069000.1 | - | 14 - 15 | 1246 | 123 | ||
More Information
Set Membership: |
|

LOC4335342
Genomic Map