Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 2 - 3 | 300 | 72 | |
| XM_015781018.2 | XP_015636504.1 | 2 - 3 | 230 | 72 | ||
| XM_015781021.2 | XP_015636507.1 | 1 - 2 | 103 | 72 | ||
| GenBank mRNA | AK069000.1 | - | 2 - 3 | 230 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015781018.2, XM_015781021.2 transcripts.
Set Membership: |
|

LOC4335342
Genomic Map