Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 3 - 4 | 336 | 79 | |
| GenBank mRNA | AK069000.1 | - | 3 - 4 | 266 | 79 | |
More Information
Set Membership: |
|

LOC4335342
Genomic Map