Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 4 - 5 | 431 | 67 | |
| XM_015781018.2 | XP_015636504.1 | 4 - 5 | 330 | 67 | ||
| GenBank mRNA | AK069000.1 | - | 4 - 5 | 361 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015781018.2 transcripts.
Set Membership: |
|

LOC4335342
Genomic Map