Assay Details
| Gene Symbol: LOC4335342 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781016.2 | XP_015636502.1 | 5 - 6 | 547 | 87 | |
| XM_015781018.2 | XP_015636504.1 | 5 - 6 | 446 | 87 | ||
| XM_015781019.2 | XP_015636505.1 | 3 - 4 | 232 | 87 | ||
| XM_015781021.2 | XP_015636507.1 | 4 - 5 | 312 | 87 | ||
| GenBank mRNA | AK069000.1 | - | 5 - 6 | 477 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015781018.2, XM_015781019.2, XM_015781021.2 transcripts.
Set Membership: |
|

LOC4335342
Genomic Map