Assay Details
Gene Symbol: LOC4335342 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015781016.2 | XP_015636502.1 | 6 - 7 | 651 | 97 | |
XM_015781018.2 | XP_015636504.1 | 6 - 7 | 550 | 97 | ||
XM_015781019.2 | XP_015636505.1 | 4 - 5 | 336 | 97 | ||
XM_015781021.2 | XP_015636507.1 | 5 - 6 | 416 | 97 | ||
GenBank mRNA | AK069000.1 | - | 6 - 7 | 581 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
