Assay Details
| Gene Symbol: LOC4335611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778074.2 | XP_015633560.1 | 2 - 3 | 268 | 148 | |
| GenBank mRNA | AK103713.1 | - | 2 - 3 | 202 | 148 | |
More Information
Set Membership: |
|

LOC4335611
Genomic Map