Assay Details
| Gene Symbol: LOC4335611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015778074.2 | XP_015633560.1 | 5 - 6 | 557 | 75 | |
| GenBank mRNA | AK061349.1 | - | 4 - 5 | 358 | 75 | |
| AK103713.1 | - | 5 - 6 | 491 | 75 | ||
More Information
Set Membership: |
|

LOC4335611
Genomic Map