Assay Details
| Gene Symbol: LOC4336216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780140.2 | XP_015635626.1 | 1 - 2 | 134 | 64 | |
| GenBank mRNA | AK064960.1 | - | 1 - 2 | 89 | 64 | |
| BT130453.1 | - | 1 - 2 | 91 | 64 | ||
| EU267971.1 | - | 1 - 2 | 72 | 64 | ||
| GQ848049.1 | - | 1 - 2 | 80 | 64 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780140.2 transcripts.
Set Membership: |
|

LOC4336216
Genomic Map