Assay Details
| Gene Symbol: LOC4336216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780140.2 | XP_015635626.1 | 2 - 3 | 157 | 77 | |
| GenBank mRNA | AK064960.1 | - | 2 - 3 | 112 | 77 | |
| BT130453.1 | - | 2 - 3 | 114 | 77 | ||
| EU267971.1 | - | 2 - 3 | 95 | 77 | ||
| GQ848049.1 | - | 2 - 3 | 103 | 77 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015780140.2 transcripts.
Set Membership: |
|

LOC4336216
Genomic Map