Assay Details
| Gene Symbol: LOC4336216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780140.2 | XP_015635626.1 | 9 - 10 | 921 | 80 | |
| GenBank mRNA | AK062122.1 | - | 5 - 6 | 483 | 80 | |
| AK064960.1 | - | 9 - 10 | 876 | 80 | ||
| BT130066.1 | - | 2 - 3 | 179 | 80 | ||
| BT131629.1 | - | 2 - 3 | 188 | 80 | ||
| EU267971.1 | - | 9 - 10 | 859 | 80 | ||
| GQ848049.1 | - | 9 - 10 | 867 | 80 | ||
More Information
Set Membership: |
|

LOC4336216
Genomic Map