Assay Details
| Gene Symbol: LOC4340394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787537.1 | XP_015643023.1 | 1 - 2 | 201 | 62 | |
| XM_015787538.1 | XP_015643024.1 | 1 - 2 | 202 | 62 | ||
| GenBank mRNA | AK060406.1 | - | 1 - 2 | 156 | 62 | |
| AK065072.1 | - | 1 - 2 | 157 | 62 | ||
| AK066195.1 | - | 1 - 2 | 157 | 62 | ||
| AK104352.1 | - | 1 - 2 | 156 | 62 | ||
| AK243612.1 | - | 1 - 2 | 157 | 62 | ||
| HQ013296.1 | - | 1 - 2 | 133 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787537.1, XM_015787538.1 transcripts.
Set Membership: |
|

LOC4340394
Genomic Map