Assay Details
| Gene Symbol: LOC4340394 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787537.1 | XP_015643023.1 | 2 - 3 | 395 | 73 | |
| XM_015787538.1 | XP_015643024.1 | 2 - 3 | 396 | 73 | ||
| GenBank mRNA | AK060406.1 | - | 2 - 3 | 350 | 73 | |
| AK065072.1 | - | 2 - 3 | 351 | 73 | ||
| AK066195.1 | - | 2 - 3 | 351 | 73 | ||
| AK104352.1 | - | 2 - 3 | 350 | 73 | ||
| AK243612.1 | - | 2 - 3 | 351 | 73 | ||
| HQ013296.1 | - | 2 - 3 | 327 | 73 | ||
More Information
Set Membership: |
|

LOC4340394
Genomic Map