Assay Details
| Gene Symbol: LOC4339495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783359.2 | XP_015638845.1 | 5 - 6 | 498 | 79 | |
| XM_015783360.2 | XP_015638846.1 | 5 - 6 | 493 | 79 | ||
More Information
Set Membership: |
|

LOC4339495
Genomic Map